siRNA Details
Gene Symbol: IL12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000882.3 | NP_000873.2 | 3 | 590 | ||
GenBank mRNA | AF101062.1 | - | 3 | 369 | ||
AF180562.1 | - | 3 | 428 | |||
BC104982.1 | - | 3 | 522 | 8143985 | ||
BC104984.1 | - | 3 | 522 | 8143987 | ||
BG702253.1 | - | 3 | 683 | |||
M65271.1 | - | 3 | 436 | |||
M65291.1 | - | 3 | 470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
