siRNA Details
Gene Symbol: IL17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002190.2 | NP_002181.1 | 3 | 1443 | ||
GenBank mRNA | U32659.1 | - | 3 | 1451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
