siRNA Details
Gene Symbol: IL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016584.2 | NP_057668.1 | 4 | 902 | ||
XM_011538477.2 | XP_011536779.1 | 6 | 1089 | |||
GenBank mRNA | AB030000.1 | - | 4 | 879 | ||
AF301620.1 | - | 4 | 886 | |||
AY359083.1 | - | 4 | 906 | |||
U38443.1 | - | 1 | 146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
