siRNA Details
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190830.1 | NP_001177759.1 | 2 | 359 | ||
NM_010589.6 | NP_034719.2 | 2 | 359 | |||
GenBank mRNA | AK038268.1 | - | 1 | 170 | ||
AK088365.1 | - | 2 | 211 | |||
AK142178.1 | - | 2 | 197 | |||
AK156646.1 | - | 2 | 181 | |||
AK164520.1 | - | 2 | 199 | |||
BC027234.1 | - | 190 | ||||
BC105577.1 | - | 1 | 162 | 3968386 | ||
BC131646.1 | - | 1 | 170 | 40110146 | ||
BC131647.1 | - | 1 | 170 | 40110148 | ||
L32955.1 | - | 1 | 161 | |||
L33768.1 | - | 1 | 155 | |||
L40172.1 | - | 2 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
