siRNA Details
Gene Symbol: Ripk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138952.3 | NP_620402.1 | 10 | 1362 | ||
GenBank mRNA | AF461040.1 | - | 10 | 1260 | ||
AF487539.1 | - | 10 | 1260 | |||
AK159593.1 | - | 10 | 1386 | |||
AK172319.1 | - | 10 | 1384 | |||
AK172417.1 | - | 10 | 1384 | |||
BC025611.1 | - | 8 | 880 | |||
BC050845.1 | - | 10 | 1376 | |||
BC058088.1 | - | 11 | 1631 | |||
BC069878.1 | - | 10 | 1387 | 5316066 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
