siRNA Details
Gene Symbol: JAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000215.3 | NP_000206.2 | 22 | 3099 | ||
XM_005259896.3 | XP_005259953.1 | 22 | 3138 | |||
GenBank mRNA | U08340.1 | - | 2 | 155 | ||
U09607.1 | - | 21 | 3094 | |||
U31601.1 | - | 22 | 3094 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
