siRNA Details
Gene Symbol: JAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000215.3 | NP_000206.2 | 11 | 1640 | ||
XM_005259896.3 | XP_005259953.1 | 11 | 1679 | |||
XM_011527990.1 | XP_011526292.1 | 11 | 1679 | |||
XM_011527991.2 | XP_011526293.1 | 11 | 1679 | |||
GenBank mRNA | AK296383.1 | - | 12 | 1755 | ||
BC028068.1 | - | 11 | 1630 | 5212575 | ||
KU178153.1 | - | 10 | 1540 | |||
KU178154.1 | - | 11 | 1654 | |||
U09607.1 | - | 10 | 1635 | |||
U31601.1 | - | 11 | 1635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
