siRNA Details
Gene Symbol: SULT1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001054.3 | NP_001045.1 | 3, 4 | 360 | ||
NM_177528.2 | NP_803564.1 | 3, 4 | 633 | |||
XM_006721077.3 | XP_006721140.1 | 2 | 1081 | |||
XM_017023600.1 | XP_016879089.1 | 3, 4 | 369 | |||
XM_017023601.1 | XP_016879090.1 | 3, 4 | 797 | |||
XM_017023602.1 | XP_016879091.1 | 4, 5 | 499 | |||
XM_017023603.1 | XP_016879092.1 | 3, 4 | 365 | |||
GenBank mRNA | BC113727.1 | - | 3, 4 | 355 | 8322779 | |
BC113729.1 | - | 3, 4 | 355 | 8322781 | ||
U28169.1 | - | 3, 4 | 355 | |||
U28170.1 | - | 3, 4 | 343 | |||
X78282.1 | - | 2, 3 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
