siRNA Details
Gene Symbol: CDK11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313896.1 | NP_001300825.1 | 18, 19 | 2396 | ||
NM_001313982.1 | NP_001300911.1 | 18, 19 | 2384 | |||
NM_024011.3 | NP_076916.2 | 18, 19 | 2387 | |||
NM_033529.3 | NP_277071.2 | 18, 19 | 2357 | |||
NR_132739.1 | 18, 19 | 2271 | ||||
NR_132740.1 | 18, 19 | 2310 | ||||
GenBank mRNA | AF067518.1 | - | 18, 19 | 2020 | ||
AF067519.1 | - | 18, 19 | 2136 | |||
AF067520.1 | - | 18, 19 | 2133 | |||
AF067521.1 | - | 18, 19 | 2106 | |||
AF067522.1 | - | 18, 19 | 2145 | |||
AF067523.1 | - | 18, 19 | 2059 | |||
AF067524.1 | - | 4, 5 | 409 | |||
BC014464.1 | - | 10, 11 | 1118 | |||
BC094827.1 | - | 10, 11 | 1151 | |||
BC110905.1 | - | 13, 14 | 1348 | 6151813 | ||
U04819.1 | - | 11, 12 | 1460 | |||
U07704.1 | - | 18, 19 | 2168 | |||
U07705.1 | - | 18, 19 | 2162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
