siRNA Details
Gene Symbol: BCAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008405.2 | NP_001008405.1 | 8 | 906 | ||
NM_018844.3 | NP_061332.2 | 8 | 1152 | |||
NR_027830.1 | 7 | 1046 | ||||
XM_006716051.2 | XP_006716114.1 | 9 | 1227 | |||
XM_011516406.2 | XP_011514708.1 | 9 | 990 | |||
XM_011516408.2 | XP_011514710.1 | 9 | 1277 | |||
GenBank mRNA | AF126020.1 | - | 8 | 862 | ||
AK289401.1 | - | 8 | 909 | |||
AK294824.1 | - | 7 | 777 | |||
BC008478.1 | - | 8 | 864 | 4294273 | ||
BG818947.1 | - | 4 | 422 | |||
BX647653.1 | - | 8 | 862 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
