siRNA Details
Gene Symbol: BCAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008405.2 | NP_001008405.1 | 4 | 387 | ||
NM_018844.3 | NP_061332.2 | 5 | 712 | |||
NR_027830.1 | 4 | 606 | ||||
XM_006716051.2 | XP_006716114.1 | 5 | 708 | |||
XM_011516406.2 | XP_011514708.1 | 5 | 471 | |||
XM_011516407.1 | XP_011514709.1 | 5 | 708 | |||
XM_011516408.2 | XP_011514710.1 | 5 | 708 | |||
XM_011516409.2 | XP_011514711.1 | 5 | 708 | |||
XM_017012436.1 | XP_016867925.1 | 5 | 708 | |||
XM_017012437.1 | XP_016867926.1 | 5 | 708 | |||
XM_017012438.1 | XP_016867927.1 | 5 | 708 | |||
GenBank mRNA | AF126020.1 | - | 5 | 422 | ||
AK289401.1 | - | 5 | 469 | |||
AK294824.1 | - | 4 | 337 | |||
AK298594.1 | - | 2 | 113 | |||
AW978976.1 | - | 1 | 32 | |||
BC008478.1 | - | 5 | 424 | 4294273 | ||
BM023571.1 | - | 2 | 182 | |||
BP261060.1 | - | 2 | 51 | |||
BT006981.1 | - | 4 | 373 | |||
BX457671.2 | - | 4 | 276 | |||
BX647653.1 | - | 5 | 422 | |||
CK820004.1 | - | 3 | 244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
