siRNA Details
Gene Symbol: BCAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008405.2 | NP_001008405.1 | 8 | 2111 | ||
NM_018844.3 | NP_061332.2 | 8 | 2357 | |||
NR_027830.1 | 7 | 2251 | ||||
XM_006716051.2 | XP_006716114.1 | 9 | 2432 | |||
XM_011516406.2 | XP_011514708.1 | 9 | 2195 | |||
XM_011516408.2 | XP_011514710.1 | 9 | 2482 | |||
GenBank mRNA | AK000878.1 | - | 1 | 540 | ||
BX647653.1 | - | 8 | 2067 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
