siRNA Details
Gene Symbol: TMEM132A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017870.3 | NP_060340.2 | 6 | 1230 | ||
NM_178031.2 | NP_821174.1 | 6 | 1227 | |||
XM_017017951.1 | XP_016873440.1 | 5 | 2827 | |||
XM_017017952.1 | XP_016873441.1 | 5 | 2823 | |||
GenBank mRNA | AB046803.1 | - | 5 | 1070 | ||
AK000546.1 | - | 4 | 447 | |||
AL359557.2 | - | 3 | 475 | |||
AL834133.1 | - | 5 | 1089 | |||
BC028106.1 | - | 6 | 1426 | |||
BC032059.1 | - | 6 | 1190 | |||
BC114569.1 | - | 6 | 1192 | 40036200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
