siRNA Details
Gene Symbol: TMEM132A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017870.3 | NP_060340.2 | 6 | 1391 | ||
NM_178031.2 | NP_821174.1 | 6 | 1388 | |||
XM_017017951.1 | XP_016873440.1 | 5 | 2988 | |||
XM_017017952.1 | XP_016873441.1 | 5 | 2984 | |||
GenBank mRNA | AB046803.1 | - | 5 | 1231 | ||
AK000546.1 | - | 4 | 608 | |||
AK023577.1 | - | 1 | 107 | |||
AL359557.2 | - | 3 | 636 | |||
AL834133.1 | - | 5 | 1250 | |||
BC028106.1 | - | 6 | 1587 | |||
BC032059.1 | - | 6 | 1351 | |||
BC114569.1 | - | 6 | 1353 | 40036200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
