siRNA Details
Gene Symbol: TMEM132A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017870.3 | NP_060340.2 | 11 | 2829 | ||
NM_178031.2 | NP_821174.1 | 11 | 2826 | |||
XM_017017951.1 | XP_016873440.1 | 10 | 4426 | |||
XM_017017952.1 | XP_016873441.1 | 10 | 4422 | |||
GenBank mRNA | AK000546.1 | - | 9 | 2046 | ||
AK023577.1 | - | 6 | 1545 | |||
BC114569.1 | - | 11 | 2791 | 40036200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
