siRNA Details
Gene Symbol: IFRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007245.2 | NP_001007246.1 | 6 | 1015 | ||
NM_001197079.1 | NP_001184008.1 | 5 | 905 | |||
NM_001197080.1 | NP_001184009.1 | 5 | 740 | |||
NM_001550.3 | NP_001541.2 | 5 | 806 | |||
XM_011516142.2 | XP_011514444.1 | 5 | 801 | |||
XM_011516143.2 | XP_011514445.1 | 5 | 986 | |||
XM_017012151.1 | XP_016867640.1 | 5 | 1053 | |||
GenBank mRNA | AK298894.1 | - | 5 | 546 | ||
AK308066.1 | - | 6 | 990 | |||
AK313211.1 | - | 6 | 1015 | |||
AK315967.1 | - | 5 | 738 | |||
BC001272.1 | - | 5 | 759 | 3458309 | ||
BT019889.1 | - | 5 | 545 | |||
BT019890.1 | - | 5 | 545 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
