siRNA Details
Gene Symbol: IFRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007245.2 | NP_001007246.1 | 13 | 1794 | ||
NM_001197079.1 | NP_001184008.1 | 12 | 1684 | |||
NM_001197080.1 | NP_001184009.1 | 12 | 1519 | |||
NM_001550.3 | NP_001541.2 | 12 | 1585 | |||
NR_120333.1 | 12 | 1441 | ||||
XM_011516142.2 | XP_011514444.1 | 13 | 1595 | |||
XM_011516143.2 | XP_011514445.1 | 12 | 1780 | |||
XM_017012151.1 | XP_016867640.1 | 12 | 1832 | |||
GenBank mRNA | AK298894.1 | - | 12 | 1325 | ||
AK313211.1 | - | 13 | 1794 | |||
AK315967.1 | - | 12 | 1517 | |||
BC001272.1 | - | 12 | 1538 | 3458309 | ||
BI915700.1 | - | 2 | 85 | |||
BT019889.1 | - | 12 | 1324 | |||
BT019890.1 | - | 12 | 1324 | |||
BX648799.1 | - | 4 | 1022 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
