siRNA Details
Gene Symbol: KIAA0100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014680.3 | NP_055495.2 | 21 | 4068 | ||
XM_005258073.2 | XP_005258130.1 | 21 | 4063 | |||
XM_017025423.1 | XP_016880912.1 | 20 | 3917 | |||
GenBank mRNA | BC120873.1 | - | 21 | 3927 | 40075102 | |
CR933634.1 | - | 22 | 3930 | |||
D43947.3 | - | 21 | 4040 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
