siRNA Details
Gene Symbol: CD79B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK222954.1 | - | 6 | 1150 | ||
AK223210.1 | - | 6 | 1155 | |||
AX775815.1 | - | 6 | 1146 | |||
AX775817.1 | - | 6 | 1143 | |||
BC002975.1 | - | 6 | 1130 | 3542354 | ||
BC032651.1 | - | 6 | 1113 | 5504755 | ||
M89957.1 | - | 6 | 1171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
