siRNA Details
Gene Symbol: CD79B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK222954.1 | - | 4 | 599 | ||
AK223210.1 | - | 4 | 604 | |||
AK313583.1 | - | 4 | 596 | |||
AX775815.1 | - | 4 | 595 | |||
AX775817.1 | - | 4 | 592 | |||
BC002975.1 | - | 4 | 579 | 3542354 | ||
BC032651.1 | - | 4 | 562 | 5504755 | ||
M80461.1 | - | 4 | 542 | |||
M89957.1 | - | 4 | 620 | |||
S52229.1 | - | 4 | 526 | |||
S79249.1 | - | 3 | 214 | |||
X83539.1 | - | 3 | 214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
