siRNA Details
Gene Symbol: CD79B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK222954.1 | - | 6 | 732 | ||
AK223210.1 | - | 6 | 737 | |||
AK313583.1 | - | 6 | 729 | |||
AX775815.1 | - | 6 | 728 | |||
AX775817.1 | - | 6 | 725 | |||
BC002975.1 | - | 6 | 712 | 3542354 | ||
BC032651.1 | - | 6 | 695 | 5504755 | ||
M80461.1 | - | 6 | 675 | |||
M89957.1 | - | 6 | 753 | |||
S52229.1 | - | 6 | 659 | |||
S79249.1 | - | 5 | 347 | |||
X83539.1 | - | 5 | 347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
