siRNA Details
Gene Symbol: MAPK8IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012324.4 | NP_036456.1 | 12 | 2560 | ||
XM_011530679.2 | XP_011528981.1 | 12 | 2452 | |||
XM_011530680.2 | XP_011528982.1 | 12 | 2338 | |||
XM_011530681.2 | XP_011528983.1 | 12 | 2317 | |||
GenBank mRNA | AF136382.1 | - | 12 | 2543 | ||
AF218778.1 | - | 10 | 2345 | |||
AK293580.1 | - | 10 | 931 | |||
AK294265.1 | - | 13 | 1619 | |||
AL021708.1 | - | 8 | 1563 | |||
BC009940.2 | - | 11 | 1310 | 4100637 | ||
BC047527.1 | - | 12 | 2473 | 5277237 | ||
U79261.1 | - | 7 | 634 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
