siRNA Details
Gene Symbol: MAPK8IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012324.4 | NP_036456.1 | 10 | 2401 | ||
XM_011530679.2 | XP_011528981.1 | 10 | 2293 | |||
XM_011530680.2 | XP_011528982.1 | 10 | 2179 | |||
XM_011530681.2 | XP_011528983.1 | 10 | 2158 | |||
GenBank mRNA | AF136382.1 | - | 10 | 2384 | ||
AF218778.1 | - | 8 | 2186 | |||
AK293580.1 | - | 8 | 772 | |||
AK294265.1 | - | 11 | 1460 | |||
AL021708.1 | - | 6 | 1404 | |||
BC009940.2 | - | 9 | 1151 | 4100637 | ||
BC047527.1 | - | 10 | 2314 | 5277237 | ||
U79261.1 | - | 5 | 475 |

More Information
Other Species Targeted

Gene Ontology Categories:
