siRNA Details
Gene Symbol: SOGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012279.2 | NP_001012279.1 | 10, 11 | 4608 | ||
GenBank mRNA | AK091416.1 | - | 7, 8 | 2379 |
Gene Symbol: KIAA0408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014702.4 | NP_055517.3 | 4, 5 | 914 | ||
GenBank mRNA | AB007868.1 | - | 2, 3 | 1307 | ||
AK091424.1 | - | 5, 6 | 1270 | |||
AK125983.1 | - | 5, 6 | 1423 | |||
BC018396.1 | - | 4, 5 | 752 | 4184033 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
