siRNA Details
Gene Symbol: KIAA0408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014702.4 | NP_055517.3 | 5 | 1040 | ||
GenBank mRNA | AK125983.1 | - | 6 | 1549 |
Gene Symbol: SOGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012279.2 | NP_001012279.1 | 11 | 4734 | ||
GenBank mRNA | AK091416.1 | - | 8 | 2505 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
