siRNA Details
Gene Symbol: LGALS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020129.2 | NP_064514.1 | 3 | 443 | ||
NM_203471.1 | NP_982297.1 | 4 | 748 | |||
GenBank mRNA | AF123762.1 | - | 2 | 220 | ||
AF267852.1 | - | 3 | 443 | |||
AF367430.1 | - | 3 | 443 | |||
AF367431.1 | - | 3 | 443 | |||
AF367432.1 | - | 3 | 443 | |||
AK312040.1 | - | 3 | 282 | |||
BC022257.1 | - | 3 | 299 | 4770140 | ||
BG434228.1 | - | 4 | 574 | |||
FJ613341.1 | - | 3 | 247 | |||
FJ613342.1 | - | 3 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
