siRNA Details
Gene Symbol: GRIPAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020137.4 | NP_064522.3 | 7 | 523 | ||
XM_011543935.1 | XP_011542237.1 | 7 | 523 | |||
XM_011543936.1 | XP_011542238.1 | 7 | 523 | |||
XM_011543937.1 | XP_011542239.1 | 7 | 570 | |||
XM_017029656.1 | XP_016885145.1 | 7 | 523 | |||
XM_017029657.1 | XP_016885146.1 | 7 | 894 | |||
XM_017029658.1 | XP_016885147.1 | 7 | 1833 | |||
XM_017029659.1 | XP_016885148.1 | 7 | 570 | |||
XM_017029660.1 | XP_016885149.1 | 7 | 924 | |||
XM_017029661.1 | XP_016885150.1 | 7 | 570 | |||
GenBank mRNA | AB032993.1 | - | 7 | 468 | ||
AK292158.1 | - | 7 | 494 | |||
BC101544.1 | - | 7 | 521 | 8069050 | ||
BC101546.1 | - | 7 | 521 | 8069052 | ||
BX640704.1 | - | 1 | 207 | |||
BX647801.1 | - | 6 | 363 | |||
BX647804.1 | - | 6 | 363 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
