siRNA Details
Gene Symbol: RANBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300865.1 | NP_001287794.1 | 9 | 834 | ||
NM_003624.2 | NP_003615.2 | 10 | 1038 | |||
NM_007320.2 | NP_015559.2 | 9 | 849 | |||
NM_007322.2 | NP_015561.1 | 10 | 1053 | |||
XM_006722928.1 | XP_006722991.1 | 11 | 1040 | |||
XM_006722929.1 | XP_006722992.1 | 9 | 676 | |||
XM_011528391.1 | XP_011526693.1 | 9 | 799 | |||
XM_011528392.1 | XP_011526694.1 | 10 | 933 | |||
XM_011528393.1 | XP_011526695.1 | 9 | 644 | |||
XM_011528394.1 | XP_011526696.1 | 7 | 505 | |||
XM_017027405.1 | XP_016882894.1 | 6 | 495 | |||
XM_017027406.1 | XP_016882895.1 | 6 | 467 | |||
GenBank mRNA | AK025300.1 | - | 10 | 889 | ||
AK125183.1 | - | 5 | 1388 | |||
AK222813.1 | - | 9 | 633 | |||
AK222990.1 | - | 10 | 837 | |||
AK299260.1 | - | 6 | 500 | |||
AK299856.1 | - | 9 | 782 | |||
AK301942.1 | - | 9 | 618 | |||
AK308205.1 | - | 10 | 765 | |||
AK314343.1 | - | 9 | 680 | |||
AL050149.1 | - | 10 | 836 | |||
BC004349.1 | - | 9 | 631 | 3350317 | ||
DQ234346.1 | - | 9 | 607 | |||
Y08697.1 | - | 8 | 614 | |||
Y08698.1 | - | 10 | 831 | |||
Y08699.1 | - | 4 | 1128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
