siRNA Details
Gene Symbol: RANBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300865.1 | NP_001287794.1 | 10 | 991 | ||
NM_003624.2 | NP_003615.2 | 11 | 1195 | |||
NM_007320.2 | NP_015559.2 | 10 | 1006 | |||
NM_007322.2 | NP_015561.1 | 11 | 1210 | |||
XM_006722928.1 | XP_006722991.1 | 12 | 1197 | |||
XM_006722929.1 | XP_006722992.1 | 10 | 833 | |||
XM_011528391.1 | XP_011526693.1 | 10 | 956 | |||
XM_011528392.1 | XP_011526694.1 | 11 | 1090 | |||
XM_011528393.1 | XP_011526695.1 | 10 | 801 | |||
XM_011528394.1 | XP_011526696.1 | 8 | 662 | |||
XM_017027405.1 | XP_016882894.1 | 7 | 652 | |||
XM_017027406.1 | XP_016882895.1 | 7 | 624 | |||
GenBank mRNA | AK025300.1 | - | 11 | 1046 | ||
AK125183.1 | - | 6 | 1545 | |||
AK222813.1 | - | 10 | 790 | |||
AK222990.1 | - | 11 | 994 | |||
AK299260.1 | - | 7 | 657 | |||
AK299856.1 | - | 10 | 939 | |||
AK301942.1 | - | 10 | 775 | |||
AK308205.1 | - | 11 | 922 | |||
AK314343.1 | - | 10 | 837 | |||
AL050149.1 | - | 11 | 993 | |||
BC004349.1 | - | 10 | 788 | 3350317 | ||
DQ234346.1 | - | 10 | 764 | |||
Y08697.1 | - | 9 | 771 | |||
Y08698.1 | - | 11 | 988 | |||
Y08699.1 | - | 4 | 2012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
