siRNA Details
Gene Symbol: UTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006786.3 | NP_006777.1 | 2 | 160 | ||
NM_021995.2 | NP_068835.1 | 3 | 224 | |||
XM_011540537.2 | XP_011538839.1 | 4 | 673 | |||
XM_011540538.1 | XP_011538840.1 | 3 | 318 | |||
XM_017000116.1 | XP_016855605.1 | 4 | 673 | |||
XM_017000117.1 | XP_016855606.1 | 3 | 318 | |||
XM_017000118.1 | XP_016855607.1 | 2 | 125 | |||
XM_017000119.1 | XP_016855608.1 | 4 | 673 | |||
GenBank mRNA | AF104118.1 | - | 2 | 151 | ||
AF140630.1 | - | 3 | 164 | |||
AY358375.1 | - | 2 | 160 | |||
BC126443.1 | - | 2 | 201 | 8992159 | ||
BC171734.1 | - | 2 | 201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
