siRNA Details
Gene Symbol: UTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006786.3 | NP_006777.1 | 4 | 434 | ||
NM_021995.2 | NP_068835.1 | 5 | 498 | |||
XM_011540537.2 | XP_011538839.1 | 6 | 947 | |||
XM_011540538.1 | XP_011538840.1 | 5 | 592 | |||
GenBank mRNA | AF104118.1 | - | 4 | 425 | ||
AY358375.1 | - | 4 | 434 | |||
BC126443.1 | - | 4 | 475 | 8992159 | ||
BC171734.1 | - | 4 | 475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
