siRNA Details
Gene Symbol: HDAC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098416.3 | NP_001091886.1 | 21 | 2571 | ||
NM_001308090.1 | NP_001295019.1 | 21 | 2631 | |||
NM_015401.4 | NP_056216.2 | 22 | 2682 | |||
XM_011538478.1 | XP_011536780.1 | 22 | 2634 | |||
XM_011538479.1 | XP_011536781.1 | 23 | 3076 | |||
XM_011538480.1 | XP_011536782.1 | 21 | 2523 | |||
XM_011538481.1 | XP_011536783.1 | 22 | 2527 | |||
XM_011538482.1 | XP_011536784.1 | 22 | 2611 | |||
XM_017019455.1 | XP_016874944.1 | 21 | 2583 | |||
GenBank mRNA | AF239243.1 | - | 19 | 2474 | ||
AK001032.1 | - | 6 | 422 | |||
AK001190.1 | - | 14 | 1604 | |||
AK024387.1 | - | 3416 | ||||
AK024469.1 | - | 17 | 2662 | |||
AK027781.1 | - | 4 | 321 | |||
AK122588.1 | - | 8 | 3641 | |||
AK128383.1 | - | 14 | 1525 | |||
AK299292.1 | - | 22 | 2597 | |||
AK301545.1 | - | 22 | 2545 | |||
AK303481.1 | - | 21 | 2537 | |||
AK309547.1 | - | 2 | 492 | |||
AL117455.1 | - | 18 | 2149 | |||
AY302468.1 | - | 22 | 2487 | |||
AY321367.1 | - | 21 | 2456 | |||
BC006453.1 | - | 11 | 1106 | |||
BC020505.2 | - | 14 | 1649 | |||
BC064840.1 | - | 21 | 2465 | 6179239 | ||
BT009771.1 | - | 12 | 1356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
