siRNA Details
Gene Symbol: HDAC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098416.3 | NP_001091886.1 | 19 | 2377 | ||
NM_001308090.1 | NP_001295019.1 | 19 | 2437 | |||
NM_015401.4 | NP_056216.2 | 20 | 2488 | |||
XM_011538478.1 | XP_011536780.1 | 20 | 2440 | |||
XM_011538479.1 | XP_011536781.1 | 21 | 2882 | |||
XM_011538480.1 | XP_011536782.1 | 19 | 2329 | |||
XM_011538481.1 | XP_011536783.1 | 20 | 2333 | |||
XM_011538482.1 | XP_011536784.1 | 20 | 2417 | |||
XM_017019455.1 | XP_016874944.1 | 19 | 2389 | |||
GenBank mRNA | AF239243.1 | - | 17 | 2280 | ||
AK001032.1 | - | 4 | 228 | |||
AK001190.1 | - | 12 | 1410 | |||
AK024469.1 | - | 15 | 2468 | |||
AK027781.1 | - | 2 | 127 | |||
AK122588.1 | - | 6 | 3447 | |||
AK128383.1 | - | 12 | 1331 | |||
AK299292.1 | - | 20 | 2403 | |||
AK301545.1 | - | 20 | 2351 | |||
AK303481.1 | - | 19 | 2343 | |||
AK309547.1 | - | 1 | 63 | |||
AL117455.1 | - | 16 | 1955 | |||
AY302468.1 | - | 20 | 2293 | |||
AY321367.1 | - | 19 | 2262 | |||
BC006453.1 | - | 9 | 912 | |||
BC020505.2 | - | 12 | 1455 | |||
BC064840.1 | - | 19 | 2271 | 6179239 | ||
BT009771.1 | - | 10 | 1162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
