siRNA Details
Gene Symbol: TBC1D25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002536.2 | NP_002527.1 | 5 | 646 | ||
GenBank mRNA | AB449912.1 | - | 5 | 535 | ||
AK293521.1 | - | 4 | 476 | |||
AK293875.1 | - | 5 | 605 | |||
AK294776.1 | - | 5 | 544 | |||
AK297924.1 | - | 2 | 51 | |||
AK298884.1 | - | 4 | 505 | |||
AK316199.1 | - | 3 | 326 | |||
BC036942.1 | - | 2 | 75 | |||
BC052609.1 | - | 4 | 436 | |||
BC082255.1 | - | 3 | 546 | |||
BC101817.1 | - | 5 | 572 | 8069323 | ||
BC101819.1 | - | 5 | 572 | 8069325 | ||
BC125088.1 | - | 5 | 561 | 40117866 | ||
BC125089.1 | - | 4 | 406 | |||
L08240.1 | - | 4 | 425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
