siRNA Details
Gene Symbol: PDCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199461.1 | NP_001186390.1 | 2 | 550 | ||
NM_001199462.1 | NP_001186391.1 | 3 | 413 | |||
NM_001199463.1 | NP_001186392.1 | 3 | 413 | |||
NM_001199464.1 | NP_001186393.1 | 3 | 413 | |||
NM_002598.3 | NP_002589.2 | 2 | 550 | |||
NM_144781.2 | NP_659005.1 | 2 | 550 | |||
XM_017010926.1 | XP_016866415.1 | 2 | 491 | |||
GenBank mRNA | AJ420535.1 | - | 2 | 296 | ||
AK297343.1 | - | 3 | 413 | |||
AW966881.1 | - | 2 | 285 | |||
AY948416.1 | - | 3 | 340 | |||
AY948417.1 | - | 3 | 340 | |||
BC008378.1 | - | 2 | 512 | 4102043 | ||
BC067759.1 | - | 2 | 426 | |||
BG721244.1 | - | 2 | 523 | |||
BX385363.2 | - | 3 | 373 | |||
DC351104.1 | - | 3 | 413 | |||
S78085.1 | - | 2 | 468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
