siRNA Details
Gene Symbol: NMRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289117.1 | NP_001276046.1 | 4 | 460 | ||
NM_170678.2 | NP_733778.1 | 4 | 445 | |||
NR_110316.1 | 3 | 205 | ||||
XM_006722725.3 | XP_006722788.1 | 3 | 371 | |||
XM_006722726.3 | XP_006722789.1 | 3 | 368 | |||
XM_017026597.1 | XP_016882086.1 | 3 | 348 | |||
GenBank mRNA | AF190819.1 | - | 3 | 228 | ||
AK001663.1 | - | 4 | 455 | |||
AK022514.1 | - | 3 | 287 | |||
AL365377.1 | - | 3 | 221 | |||
AY611481.1 | - | 3 | 155 | |||
BC093637.1 | - | 3 | 220 | 7939482 | ||
BC101575.1 | - | 3 | 220 | 8069081 | ||
BC143329.1 | - | 3 | 235 | 9051834 | ||
BM717225.1 | - | 3 | 192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
