siRNA Details
Gene Symbol: FAM135A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162529.1 | NP_001156001.1 | 13 | 1451 | ||
XM_005248743.1 | XP_005248800.1 | 16 | 1832 | |||
XM_005248744.4 | XP_005248801.1 | 15 | 1799 | |||
XM_005248745.2 | XP_005248802.1 | 15 | 1754 | |||
XM_005248749.1 | XP_005248806.1 | 16 | 1832 | |||
XM_011535993.2 | XP_011534295.1 | 15 | 1552 | |||
XM_011535994.2 | XP_011534296.1 | 16 | 1972 | |||
XM_011535995.2 | XP_011534297.1 | 16 | 1773 | |||
XM_011535996.2 | XP_011534298.1 | 16 | 1930 | |||
XM_011535997.2 | XP_011534299.1 | 14 | 1674 | |||
XM_017011124.1 | XP_016866613.1 | 15 | 1805 | |||
XM_017011125.1 | XP_016866614.1 | 14 | 1577 | |||
XM_017011126.1 | XP_016866615.1 | 15 | 1891 | |||
XM_017011127.1 | XP_016866616.1 | 14 | 1673 | |||
XM_017011128.1 | XP_016866617.1 | 13 | 1597 | |||
XM_017011135.1 | XP_016866624.1 | 14 | 1653 | |||
XM_017011136.1 | XP_016866625.1 | 5 | 380 | |||
GenBank mRNA | AB037832.1 | - | 13 | 1419 | ||
AK000183.1 | - | 12 | 1585 | |||
AK225230.1 | - | 12 | 1585 | |||
AL136820.1 | - | 2 | 76 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
