siRNA Details
Gene Symbol: ZNF324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014347.2 | NP_055162.1 | 4 | 2846 | ||
XM_005258713.4 | XP_005258770.1 | 4 | 3641 | |||
GenBank mRNA | AF060503.1 | - | 4 | 2795 | ||
AF075117.1 | - | 1 | 363 | |||
AK023989.1 | - | 2 | 2109 | |||
AK092341.1 | - | 4 | 3197 | |||
BC007717.2 | - | 5 | 2686 | 4303415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
