siRNA Details
Gene Symbol: KLHL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020782.1 | NP_065833.1 | 3 | 3717 | ||
XM_017019698.1 | XP_016875187.1 | 3 | 4482 | |||
GenBank mRNA | AB037761.1 | - | 3 | 3448 | ||
BC137116.1 | - | 3 | 3684 | 9021113 | ||
BC137117.1 | - | 3 | 3683 | 9021114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
