siRNA Details
Gene Symbol: MICAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546137.2 | XP_011544439.1 | 17 | 2455 | ||
XM_011546139.2 | XP_011544441.1 | 17 | 2455 | |||
XM_011546142.2 | XP_011544444.1 | 17 | 2455 | |||
XM_011546146.2 | XP_011544448.1 | 17 | 2455 | |||
XM_017028865.1 | XP_016884354.1 | 17 | 2973 | |||
XM_017028866.1 | XP_016884355.1 | 17 | 2779 | |||
XM_017028867.1 | XP_016884356.1 | 17 | 3896 | |||
XM_017028868.1 | XP_016884357.1 | 17 | 2625 | |||
XM_017028869.1 | XP_016884358.1 | 17 | 2370 | |||
XM_017028870.1 | XP_016884359.1 | 17 | 2371 | |||
XM_017028871.1 | XP_016884360.1 | 17 | 2455 | |||
XM_017028872.1 | XP_016884361.1 | 17 | 2455 | |||
XM_017028873.1 | XP_016884362.1 | 17 | 2455 | |||
XM_017028875.1 | XP_016884364.1 | 17 | 2455 | |||
XM_017028876.1 | XP_016884365.1 | 17 | 2455 | |||
XM_017028877.1 | XP_016884366.1 | 17 | 2455 | |||
XM_017028878.1 | XP_016884367.1 | 17 | 2455 | |||
XM_017028883.1 | XP_016884372.1 | 16 | 2118 | |||
GenBank mRNA | AB037785.1 | - | 14 | 1777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
