siRNA Details
Gene Symbol: KRT23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282433.1 | NP_001269362.1 | 7 | 1283 | ||
NM_015515.4 | NP_056330.3 | 8 | 1811 | |||
XM_005257200.4 | XP_005257257.1 | 7 | 864 | |||
XM_011524595.1 | XP_011522897.1 | 7 | 854 | |||
GenBank mRNA | AF102848.1 | - | 7 | 1239 | ||
AK002047.1 | - | 7 | 1223 | |||
AK225866.1 | - | 7 | 1287 | |||
AK289449.1 | - | 7 | 1306 | |||
AK302109.1 | - | 7 | 1092 | |||
AK314604.1 | - | 7 | 1023 | |||
AL117538.1 | - | 6 | 1067 | |||
BC028356.2 | - | 8 | 1591 | 4838347 | ||
CV373638.1 | - | 4 | 499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
