siRNA Details
Gene Symbol: ERMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024896.2 | NP_079172.2 | 15 | 2710 | ||
XM_005251587.1 | XP_005251644.1 | 13 | 2315 | |||
XM_017015139.1 | XP_016870628.1 | 16 | 2503 | |||
GenBank mRNA | AB058718.2 | - | 15 | 2587 | ||
AK026962.1 | - | 7 | 1016 | |||
AK093217.1 | - | 13 | 1886 | |||
AK127218.1 | - | 15 | 2699 | |||
AK172831.1 | - | 10 | 1463 | |||
AL834191.1 | - | 14 | 2342 | |||
BC031630.1 | - | 7 | 1074 | |||
BC136771.1 | - | 15 | 2684 | 9020761 | ||
BC136773.1 | - | 15 | 2684 | 9020763 | ||
BC144404.1 | - | 16 | 2733 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
