siRNA Details
Gene Symbol: ERMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024896.2 | NP_079172.2 | 12 | 2260 | ||
XM_005251587.1 | XP_005251644.1 | 10 | 1865 | |||
XM_017015139.1 | XP_016870628.1 | 13 | 2053 | |||
GenBank mRNA | AB058718.2 | - | 12 | 2137 | ||
AK026962.1 | - | 4 | 566 | |||
AK093217.1 | - | 10 | 1436 | |||
AK127218.1 | - | 12 | 2249 | |||
AK172831.1 | - | 6 | 964 | |||
AL834191.1 | - | 11 | 1892 | |||
BC031630.1 | - | 4 | 624 | |||
BC136771.1 | - | 12 | 2234 | 9020761 | ||
BC136773.1 | - | 12 | 2234 | 9020763 | ||
BC144404.1 | - | 12 | 2234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
