siRNA Details
Gene Symbol: MZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267033.1 | NP_001253962.1 | 6 | 2610 | ||
NM_003422.2 | NP_003413.2 | 6 | 2536 | |||
NM_198055.1 | NP_932172.1 | 6 | 2854 | |||
XM_005259204.3 | XP_005259261.1 | 7 | 3209 | |||
XM_017027206.1 | XP_016882695.1 | 6 | 3057 | |||
GenBank mRNA | AF055077.1 | - | 6 | 2536 | ||
AF055078.1 | - | 6 | 2854 | |||
BC007777.2 | - | 6 | 2512 | 4301723 | ||
BC053316.1 | - | 6 | 2333 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
