siRNA Details
Gene Symbol: POP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015918.3 | NP_057002.2 | 3, 4 | 353 | ||
GenBank mRNA | AF070660.1 | - | 3, 4 | 337 | ||
AF117232.1 | - | 3, 4 | 337 | |||
AJ306296.1 | - | 3, 4 | 340 | |||
AK223206.1 | - | 3, 4 | 325 | |||
BC012505.1 | - | 3, 4 | 340 | 4708484 | ||
BI668578.1 | - | 3, 4 | 360 | |||
CB995722.1 | - | 3, 4 | 475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
