siRNA Details
Gene Symbol: TECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321170.1 | NP_001308099.1 | 7 | 560 | ||
NM_138501.5 | NP_612510.1 | 6 | 486 | |||
NR_038103.1 | 6 | 518 | ||||
NR_038104.1 | 6 | 551 | ||||
XM_006722945.1 | XP_006723008.1 | 7 | 604 | |||
XM_011528442.2 | XP_011526744.1 | 6 | 434 | |||
GenBank mRNA | AF038958.1 | - | 6 | 432 | ||
AF222742.1 | - | 6 | 407 | |||
AK001416.1 | - | 6 | 450 | |||
AK094087.1 | - | 7 | 527 | |||
AK299134.1 | - | 7 | 524 | |||
AK315412.1 | - | 6 | 450 | |||
BC000174.2 | - | 5 | 346 | |||
BC002425.1 | - | 6 | 418 | 3163592 | ||
BC005952.1 | - | 6 | 429 | 4249841 | ||
BC007801.2 | - | 6 | 436 | 4128702 | ||
BC013881.2 | - | 6 | 434 | 3928698 | ||
BG705196.1 | - | 6 | 489 | |||
BT007179.1 | - | 5 | 356 | |||
CR456892.1 | - | 5 | 356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
