siRNA Details
Gene Symbol: SF3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005877.5 | NP_005868.1 | 16 | 2776 | ||
GenBank mRNA | AK098463.1 | - | 9 | 1542 | ||
AK303273.1 | - | 17 | 2701 | |||
BC001976.1 | - | 16 | 2716 | 3459964 | ||
BC007684.2 | - | 16 | 2743 | 3634035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
