siRNA Details
Gene Symbol: SF3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005877.5 | NP_005868.1 | 3 | 361 | ||
GenBank mRNA | AK303273.1 | - | 3 | 336 | ||
BC001976.1 | - | 3 | 301 | 3459964 | ||
BC007684.2 | - | 3 | 328 | 3634035 | ||
BI827130.1 | - | 3 | 315 | |||
CR456575.1 | - | 3 | 304 | |||
DB445415.1 | - | 3 | 361 | |||
X85237.1 | - | 3 | 303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
