siRNA Details
Gene Symbol: MORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303256.2 | NP_001290185.1 | 23, 24 | 3145 | ||
NM_001303257.2 | NP_001290186.1 | 23, 24 | 3145 | |||
NM_014941.3 | NP_055756.1 | 24, 25 | 4011 | |||
XM_011530004.1 | XP_011528306.1 | 23, 24 | 3370 | |||
XM_017028667.1 | XP_016884156.1 | 23, 24 | 3370 | |||
XM_017028668.1 | XP_016884157.1 | 24, 25 | 3465 | |||
GenBank mRNA | AB020659.1 | - | 24, 25 | 3925 | ||
AL133637.1 | - | 5, 6 | 616 | |||
BC019257.1 | - | 22, 23 | 2672 | 3503651 | ||
BC136782.1 | - | 24, 25 | 2870 | 9020772 | ||
CR456469.1 | - | 22, 23 | 2671 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
