siRNA Details
Gene Symbol: MORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303256.2 | NP_001290185.1 | 26 | 3597 | ||
NM_001303257.2 | NP_001290186.1 | 26 | 3588 | |||
NM_014941.3 | NP_055756.1 | 27 | 4463 | |||
XM_011530004.1 | XP_011528306.1 | 26 | 3822 | |||
XM_017028667.1 | XP_016884156.1 | 26 | 3813 | |||
XM_017028668.1 | XP_016884157.1 | 27 | 3917 | |||
GenBank mRNA | AB020659.1 | - | 27 | 4377 | ||
AL133637.1 | - | 8 | 1068 | |||
BC019257.1 | - | 25 | 3124 | 3503651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
